missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-2-methyl-1-propanol 93.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Boiling Point | 165°C |
| Quantity | 500 mL |
| Formula Weight | 89.14 |
| Percent Purity | ≥93.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 2-Amino-2-methyl-1-propanol |
Safety and Handling
| EINECSNumber | (9)-0118 |
| RTECSNumber | UA5950000 |
| TSCA | Yes |