missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Amino-2-methyl-1-propanol

Catalog No. A651821L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A651821L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A651821L Supplier Sigma Aldrich Supplier No. A651821L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Amino-2-methyl-1-propanol

Specifications

CAS 124-68-5
Melting Point 24°C to 28°C (lit.)
Density 0.934 g/mL (at 25°C (literature))
Boiling Point 165°C (lit.)
Molecular Formula C4H11NO
Refractive Index n20/D 1.4455 (literature)
Linear Formula (CH3)2 C(NH2)CH2OH
MDL Number MFCD00008051
Quantity 1 L
Synonym beta-Aminoisobutyl alcohol; AMP; AMP 95
Molecular Weight (g/mol) 89.14
Percent Purity 90%
Show More Show Less