accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOH

CAS RN 123855-51-6

IUPAC Name: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 215.29
Molecular Formula: C11H21NO3
InChi Key: CTEDVGRUGMPBHE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CO)CC1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)