accessibility menu, dialog, popup

UserName

OSOOOOV

CAS RN 123334-20-3

IUPAC Name: oxovanadiumbis(ylium) sulfate
Synonyms: vanadyl sulfate|vanadium oxysulfate|vanadic sulfate|vanadium oxide sulphate|oxo[sulfato(2-)-O]-vanadium|vanadin(IV) oxide sulfate
Molecular Weight (g/mol): 163
Molecular Formula: O5SV
InChi Key: UUUGYDOQQLOJQA-UHFFFAOYSA-L
SMILES: O=[V++].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (5)
Grade 
  • (5)
Molecular Weight (g/mol) 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (5)
  • (1)
  • (3)