missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Vanadium(IV) oxide sulfate hydrate
Description
Vanadium(IV) oxide sulfate hydrate
Specifications
Specifications
| Melting Point | 105°C |
| CAS | 123334-20-3 |
| Quantity | 500 g |
| Molecular Formula | O5SV -+ xH2O |
| Linear Formula | VOSO4 -+ xH2O |
| MDL Number | MFCD00149933 |
| Synonym | Vanadyl sulfate hydrate |
| Molecular Weight (g/mol) | 163 |
| Percent Purity | 97% |