accessibility menu, dialog, popup

UserName

CAS RN 123-92-2

IUPAC Name: 3-methylbutyl acetate
Synonyms: banana oil|pear oil|I-amyl acetate|isoamylacetat|isoamyl acetate (natural)|amylacetic ester|amyl acetate, common|acetate d'isoamyle|fema number 2055|isoamylester kyseliny octove
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: MLFHJEHSLIIPHL-UHFFFAOYSA-N
SMILES: CC(C)CCOC(C)=O
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)