missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl acetate, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020619
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020619 1 mL
111019345 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020619 Supplier Merck Emd Millipore Supplier No. 798571ML

Isopentyl acetate is an alarm pheromone present in honey bees, which may affect the brain and hence, provoke a quick defensive response, thereby influencing their behavior for a longer period of time.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 680°F
CAS 123-92-2
Vapor Pressure 5 mmHg (25°C)
Vapor Density 4.5 (vs. air)
Density 0.876 g/mL (at 25°C)
Flash Point 33°C - Closed Cup
Physical Form Neat
Linear Formula CH3 COOCH2 CH2 CH(CH3)2
MDL Number MFCD00008946
Melting Point -78°C
Boiling Point 142°C (756 mmHg)
Synonym Acetic acid 3-methylbutyl ester; Isoamyl acetate
Type Analytical Standard
Quantity 1 mL
Formula Weight 130.18
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less