accessibility menu, dialog, popup

UserName

NH2OH

CAS RN 123-30-8

IUPAC Name: 4-aminophenol
Synonyms: aminophenol renal AC 4-aminophenol, 18o-labeled Unal benzofur P 3h-labeled 4-aminophenol 4-aminophenol, 3h-labeled C.I. oxidation base 6 paranol paramidophenol
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: PLIKAWJENQZMHA-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C=C1
Boiling Point 
  • (2)
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (8)
  • (2)
  • (4)
  • (4)
  • (6)
  • (4)
  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)