missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenol 98.0+%, TCI America™

Catalog No. A0384500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0384500G 500 g
A038425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0384500G Supplier TCI America Supplier No. A0384500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-30-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007869
InChI Key PLIKAWJENQZMHA-UHFFFAOYSA-N
Synonym p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PubChem CID 403
ChEBI CHEBI:17602
IUPAC Name 4-aminophenol
SMILES C1=CC(=CC=C1N)O

Specifications

Melting Point 187°C
Boiling Point 284°C
Quantity 500 g
UN Number 2512
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenol
EINECSNumber (3)-0675
RTECSNumber SJ5075000
TSCA Yes
Recommended Storage Refrigerator