accessibility menu, dialog, popup

UserName

CAS RN 122852-69-1

IUPAC Name: 5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-1H,2H,3H,4H,5H-pyrido[4,3-b]indol-1-one hydrochloride
Synonyms: alosetron hydrochloride
Molecular Weight (g/mol): 330.82
Molecular Formula: C17H19ClN4O
InChi Key: FNYQZOVOVDSGJH-UHFFFAOYSA-N
SMILES: Cl.CN1C2=C(C3=CC=CC=C13)C(=O)N(CC1=C(C)NC=N1)CC2
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)