missing translation for 'onlineSavingsMsg'
Learn More

Alosetron Hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111002504
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002504 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002504 Supplier Merck Emd Millipore Supplier No. 1013807200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 122852-69-1
Type USP Reference Standard
Quantity 200 mg
Grade Pharmaceutical Primary Standard