accessibility menu, dialog, popup

UserName

CAS RN 1226-42-2

IUPAC Name: bis(4-methoxyphenyl)ethane-1,2-dione
Synonyms: bis(4-methoxyphenyl)ethane-1,2-dione
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: YNANGXWUZWWFKX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)