missing translation for 'onlineSavingsMsg'
Learn More

p-Anisil 98.0+%, TCI America™

Catalog No. A10285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10285G 5 g
A102825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10285G Supplier TCI America Supplier No. A10285G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1226-42-2
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00008405
InChI Key YNANGXWUZWWFKX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
PubChem CID 71043
IUPAC Name 1,2-bis(4-methoxyphenyl)ethane-1,2-dione
SMILES COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 270.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Anisil
EINECSNumber (4)-0277
TSCA Yes