accessibility menu, dialog, popup

UserName

CAS RN 1225023-87-9

IUPAC Name: 3-[1-(2-carboxyethyl)-16-[(4-nitrophenyl)carbamoyl]-15-oxa-16-azatetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-8-yl]propanoic acid
Synonyms: 3-[1-(2-carboxyethyl)-16-[(4-nitrophenyl)carbamoyl]-15-oxa-16-azatetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-8-yl]propanoic acid
Molecular Weight (g/mol): 517.49
Molecular Formula: C27H23N3O8
InChi Key: IGXYVYDHRNQRAA-UHFFFAOYSA-N
SMILES: OC(=O)CCC12ON(C(=O)NC3=CC=C(C=C3)[N+]([O-])=O)C(CCC(O)=O)(C3=CC=CC=C13)C1=CC=CC=C21
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results