missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydro-9,10-bis(2-carboxyethyl)-N-(4-nitrophenyl)-10,9-(epoxyimino)anthracene-12-carboxamide, TCI America™

Catalog No. D3929100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3929100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3929100MG Supplier TCI America Supplier No. D3929100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1225023-87-9
Molecular Formula C27H23N3O8
Molecular Weight (g/mol) 517.49
MDL Number MFCD15072162
InChI Key IGXYVYDHRNQRAA-UHFFFAOYSA-N
PubChem CID 44630026
IUPAC Name 3-[1-(2-carboxyethyl)-16-[(4-nitrophenyl)carbamoyl]-15-oxa-16-azatetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-8-yl]propanoic acid
SMILES OC(=O)CCC12ON(C(=O)NC3=CC=C(C=C3)[N+]([O-])=O)C(CCC(O)=O)(C3=CC=CC=C13)C1=CC=CC=C21

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 517.49
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dihydro-9,10-bis(2-carboxyethyl)-N-(4-nitrophenyl)-10,9-(epoxyimino)anthracene-12-carboxamide
TSCA No
Recommended Storage Refrigerator