accessibility menu, dialog, popup

UserName

CAS RN 122111-01-7

IUPAC Name: [3-(benzoyloxy)-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate
Synonyms: [3-(benzoyloxy)-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate
Molecular Weight (g/mol): 376.31
Molecular Formula: C19H14F2O6
InChi Key: SHHNEUNVMZNOID-UHFFFAOYNA-N
SMILES: FC1(F)C(OC(=O)C2=CC=CC=C2)C(COC(=O)C2=CC=CC=C2)OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results