missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-2,2-difluoro-D-erythro-pentonic Acid gamma-Lactone 3,5-Dibenzoate 98.0+%, TCI America™

Catalog No. D42075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42075G 5 g
D42071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42075G Supplier TCI America Supplier No. D42075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122111-01-7
Molecular Formula C19H14F2O6
Molecular Weight (g/mol) 376.312
MDL Number MFCD08458308
InChI Key SHHNEUNVMZNOID-HUUCEWRRSA-N
PubChem CID 9821015
IUPAC Name [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(=O)O2)(F)F)OC(=O)C3=CC=CC=C3

Specifications

Melting Point 123°C
Color White
Quantity 5 g
Formula Weight 376.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Deoxy-2,2-difluoro-D-erythro-pentonic Acid gamma-Lactone 3,5-Dibenzoate
TSCA No
Recommended Storage Freezer