accessibility menu, dialog, popup

UserName

CH3OPOH3COCH3

CAS RN 122-52-1

IUPAC Name: triethyl phosphite
Synonyms: triethyl phosphite
Molecular Weight (g/mol): 166.16
Molecular Formula: C6H15O3P
InChi Key: BDZBKCUKTQZUTL-UHFFFAOYSA-N
SMILES: CCOP(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)