accessibility menu, dialog, popup

UserName

CAS RN 1219-99-4

IUPAC Name: 1,3-bis(4-chlorophenyl)urea
Synonyms: 1,3-bis(4-chlorophenyl)urea
Molecular Weight (g/mol): 281.14
Molecular Formula: C13H10Cl2N2O
InChi Key: ZNQCSLYENQIUMJ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(NC(=O)NC2=CC=C(Cl)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results