accessibility menu, dialog, popup

UserName

CAS RN 121899-81-8

IUPAC Name: dichlorotris(2-methoxyphenyl)bismuthane
Synonyms: dichlorotris(2-methoxyphenyl)bismuthane
Molecular Weight (g/mol): 601.28
Molecular Formula: C21H21BiCl2O3
InChi Key: VWHUHQRLABAFKC-UHFFFAOYSA-L
SMILES: COC1=CC=CC=C1[Bi](Cl)(Cl)(C1=CC=CC=C1OC)C1=CC=CC=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results