missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methoxyphenyl)bismuth Dichloride 98.0+%, TCI America™

Catalog No. T19561G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19561G Supplier TCI America Supplier No. T19561G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121899-81-8
Molecular Formula C21H21BiCl2O3
Molecular Weight (g/mol) 601.276
MDL Number MFCD04038429
InChI Key VWHUHQRLABAFKC-UHFFFAOYSA-L
Synonym Dichlorotris(2-methoxyphenyl)bismuth
PubChem CID 10603660
IUPAC Name dichloro-tris(2-methoxyphenyl)bismuth
SMILES COC1=CC=CC=C1[Bi](C2=CC=CC=C2OC)(C3=CC=CC=C3OC)(Cl)Cl

Specifications

Melting Point 183°C
Color Yellow
Quantity 1 g
Formula Weight 601.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-methoxyphenyl)bismuth Dichloride
TSCA No
Recommended Storage Refrigerator