accessibility menu, dialog, popup

UserName

CAS RN 121859-57-2

HNClCl

CAS RN 121859-57-2

IUPAC Name: 5,6-dichloro-1H-indole
Synonyms: 5,6-dichloro-1H-indole
Molecular Weight (g/mol): 186.04
Molecular Formula: C8H5Cl2N
InChi Key: ILINOHVVKWYAFM-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=C2C=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results