CAS RN 1217503-60-0
IUPAC Name:
1-(4-hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)penta-1,4-dien-3-one
Synonyms:
1-(4-hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)penta-1,4-dien-3-one
Molecular Weight (g/mol):
400.43
Molecular Formula:
C22H24O7
InChi Key:
SKZNJIJSGSQNJQ-UHFFFAOYSA-N
SMILES:
COC1=CC(C=CC(=O)C=CC2=CC(OC)=C(OC)C(OC)=C2)=CC(OC)=C1O
1
–
1
of
1
results
1
(1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one 95.0+%, TCI America™
CAS: 1217503-60-0 Molecular Formula: C22H24O7 Molecular Weight (g/mol): 400.427 InChI Key: SKZNJIJSGSQNJQ-CDJQDVQCSA-N Synonym: GO-Y078 PubChem CID: 46231908 IUPAC Name: (1E,4E)-1-(4-hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)penta-1,4-dien-3-one SMILES: COC1=CC(=CC(=C1O)OC)C=CC(=O)C=CC2=CC(=C(C(=C2)OC)OC)OC