missing translation for 'onlineSavingsMsg'
Learn More

(1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one 95.0+%, TCI America™

Catalog No. H152520MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H152520MG 20 mg
H1525100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H152520MG Supplier TCI America Supplier No. H152520MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1217503-60-0
Molecular Formula C22H24O7
Molecular Weight (g/mol) 400.427
InChI Key SKZNJIJSGSQNJQ-CDJQDVQCSA-N
Synonym GO-Y078
PubChem CID 46231908
IUPAC Name (1E,4E)-1-(4-hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)penta-1,4-dien-3-one
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)C=CC2=CC(=C(C(=C2)OC)OC)OC

Specifications

Quantity 20 mg
Formula Weight 400.43
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one