accessibility menu, dialog, popup

UserName

CAS RN 121399-88-0

NNHNFFFFFFFFFFFFFFFFFFFFFNHFFFFFFF

CAS RN 121399-88-0

IUPAC Name: 4,5,9,10,14,15,19,20-octafluoro-2,7,12,17-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Synonyms: 4,5,9,10,14,15,19,20-octafluoro-2,7,12,17-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular Weight (g/mol): 1118.48
Molecular Formula: C44H2F28N4
InChi Key: BVYJQIBTXOTRJZ-UHFFFAOYSA-N
SMILES: FC1=C2NC(=C1F)C(=C1N=C(C(F)=C1F)C(=C1N=C(C(F)=C1F)C(=C1NC(C(F)=C1F)=C2C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results