missing translation for 'onlineSavingsMsg'
Learn More

2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin 98.0+%, TCI America™

Catalog No. O0319100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0319100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0319100MG Supplier TCI America Supplier No. O0319100MG
Only null left

Chemical Identifiers

CAS 121399-88-0
Molecular Formula C44H2F28N4
Molecular Weight (g/mol) 1118.483
InChI Key BVYJQIBTXOTRJZ-UHFFFAOYSA-N
Synonym Perfluoro-5,10,15,20-tetraphenyl-21H,23H-porphyrin, 2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)-21H,23H-porphine
PubChem CID 9898400
IUPAC Name 2,3,7,8,12,13,17,18-octafluoro-5,10,15,20-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22-dihydroporphyrin
SMILES C1(=C2C(=C(C(=C(C3=NC(=C(C4=NC(=C(C5=C(C(=C1N5)F)F)C6=C(C(=C(C(=C6F)F)F)F)F)C(=C4F)F)C7=C(C(=C(C(=C7F)F)F)F)F)C(=C3F)F)C8=C(C(=C(C(=C8F)F)F)F)F)N2)F)F)C9=C(C(=C(C(=C9F)F)F)F)F

Specifications

Quantity 100 mg
Formula Weight 1118.48
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin