CAS RN 12135-76-1
Ammonium sulfide, 40-44% w/w aq. soln.
CAS: 12135-76-1 Molecular Formula: H8N2S Molecular Weight (g/mol): 68.14 MDL Number: MFCD00010892 InChI Key: UXXGWUQNRMPNKH-UHFFFAOYSA-N IUPAC Name: diamine sulfane SMILES: N.N.S
Ammonium sulfide, 20-24% aq. soln.
CAS: 12135-76-1 Molecular Formula: H8N2S Molecular Weight (g/mol): 68.14 MDL Number: MFCD00010892 InChI Key: UXXGWUQNRMPNKH-UHFFFAOYSA-N IUPAC Name: diamine sulfane SMILES: N.N.S
Ammonium sulfide, 20% solution in water
CAS: 12135-76-1 Molecular Formula: H8N2S Molecular Weight (g/mol): 68.14 MDL Number: MFCD00010892 InChI Key: UXXGWUQNRMPNKH-UHFFFAOYSA-N IUPAC Name: diamine sulfane SMILES: N.N.S
Ammonium Sulfide 20% MP Biomedicals
CAS: 12135-76-1 Molecular Formula: H8N2S Molecular Weight (g/mol): 68.14 MDL Number: MFCD00010892 InChI Key: UXXGWUQNRMPNKH-UHFFFAOYSA-N IUPAC Name: diamine sulfane SMILES: N.N.S
Ammonium sulfide, 21.0%, MP Biomedicals™
CAS: 12135-76-1 Molecular Formula: H8N2S Molecular Weight (g/mol): 68.14 MDL Number: MFCD00010892 InChI Key: UXXGWUQNRMPNKH-UHFFFAOYSA-N IUPAC Name: diamine sulfane SMILES: N.N.S
Ammonium Sulfide, Solution, 20%, Spectrum™ Chemical
CAS: 12135-76-1
Ammonium Sulfide Solution, Reagent, ACS, 20-24%, Spectrum™ Chemical
CAS: 12135-76-1 Molecular Formula: H8N2S Molecular Weight (g/mol): 68.14 MDL Number: MFCD00010892 InChI Key: UXXGWUQNRMPNKH-UHFFFAOYSA-N IUPAC Name: diamine sulfane SMILES: N.N.S
Ammonium Sulfide, Light Solution, Reagent, 20-24%, Spectrum™ Chemical
CAS: 12135-76-1 Molecular Formula: H8N2S Molecular Weight (g/mol): 68.14 MDL Number: MFCD00010892 InChI Key: UXXGWUQNRMPNKH-UHFFFAOYSA-N IUPAC Name: diamine sulfane SMILES: N.N.S