missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 21.0%, MP Biomedicals™

Catalog No. ICN19386080
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19386080 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19386080 Supplier MP Biomedicals Inc Supplier No. 0219386080
Only null left

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Color Yellow
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 0.21
Formula Weight 68.1
Density 0.997g/mL
Chemical Name or Material Ammonium sulfide