missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfide 20% MP Biomedicals

Catalog No. ICN19386025
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19386025 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19386025 Supplier MP Biomedicals Inc Supplier No. 0219386025
Only null left

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Quantity 25 mL
Assay Percent Range 20%
Formula Weight 68.1
Density 0.997 g/mL
Chemical Name or Material Ammonium Sulfide