missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfide, 20% solution in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Quantity | 100 mL |
| Linear Formula | (NH4)2S |
| Flash Point | 32°C |
| Merck Index | 15, 553 |
| Solubility Information | Solubility in water: miscible |
| Formula Weight | 68.14 |
| Concentration or Composition (by Analyte or Components) | 20.0 to 24.0% (Iodimetry) |
| Packaging | Glass bottle |
| Appearance | Clear yellow solution |
| Specific Gravity | 1 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Flammable liquid and vapor. Very toxic to aquatic life. Contact with acids liberates toxic gas. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
| EINECSNumber | 235-223-4 |
RUO – Research Use Only