missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 20-24% aq. soln.

Catalog No. AA33286AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33286AP 500 mL
AA33286AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33286AP Supplier Thermo Scientific Chemicals Supplier No. 033286AP
Only null left

CAS: 12135-76-1 | H8N2S | 68.14 g/mol

Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth. It is an unstable salt and acts as a selective reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Color Colorless to Orange
Physical Form Liquid
Quantity 500 mL
Assay Percent Range 20 to 24% (aqueous)
Flash Point 32°C (90°F)
Solubility Information Miscible with water,alcohol and liquid ammonia.
Formula Weight 68.14
Odor Like rotten eggs
Appearance Colorless liquid
Vapor Pressure 546 hPa (410mm Hg) at 20°C
Density 0.997 g/cm3 at 20°C
Chemical Name or Material Ammonium sulfide
Show More Show Less
Hazard Category H314-H335
Hazard Statement When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Use appropriate containment to avoid environmental contamination.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packing Group: II
EINECSNumber 235-223-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only