accessibility menu, dialog, popup

UserName

CAS RN 121309-88-4

IUPAC Name: benzyltrimethylazanium; tetrachloro-λ³-iodanuide
Synonyms: benzyltrimethylazanium; tetrachloro-λ³-iodanuide
Molecular Weight (g/mol): 418.95
Molecular Formula: C10H16Cl4IN
InChi Key: IBXYFQYYVRYALP-UHFFFAOYSA-N
SMILES: Cl[I-](Cl)(Cl)Cl.C[N+](C)(C)CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results