missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Tetrachloroiodate 95.0+%, TCI America™

Catalog No. B15435G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15435G Supplier TCI America Supplier No. B15435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121309-88-4
Molecular Formula C10H16Cl4IN
Molecular Weight (g/mol) 418.949
MDL Number MFCD00191607
InChI Key IBXYFQYYVRYALP-UHFFFAOYSA-N
PubChem CID 11122622
SMILES C[N+](C)(C)CC1=CC=CC=C1.Cl[I-](Cl)(Cl)Cl

Specifications

Melting Point 135°C
Quantity 5 g
UN Number 1759
Formula Weight 418.95
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltrimethylammonium Tetrachloroiodate [Chlorinating Reagent]
EINECSNumber (3)-2694
TSCA No