accessibility menu, dialog, popup

UserName

CAS RN 121124-97-8

IUPAC Name: (3-formyl-4-methoxyphenyl)boronic acid
Synonyms: 3-formyl-4-methoxyphenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: YJQDBKGGRPJSOI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C=O)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results