missing translation for 'onlineSavingsMsg'
Learn More

3-Formyl-4-methoxybenzeneboronic acid, 98%

Catalog No. AAL1785006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1785006 5 g
AAL1785003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1785006 Supplier Thermo Scientific Chemicals Supplier No. L1785006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121124-97-8 | C8H9BO4 | 179.966 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121124-97-8
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.966
MDL Number MFCD02093661
InChI Key YJQDBKGGRPJSOI-UHFFFAOYSA-N
Synonym 3-formyl-4-methoxybenzeneboronic acid, 3-formyl-4-methoxyphenyl boronic acid, 4-borono-2-formylanisole, 5-borono-2-methoxybenzaldehyde, 5-borono-o-anisaldehyde, 3-formyl-4-methoxylphenylboronic acid, boronic acid, 3-formyl-4-methoxyphenyl, 5-dihydroxyboryl-2-methoxybenzaldehyde, 3-formyl-4-methoxy-phenyl boronic acid
PubChem CID 2759225
IUPAC Name (3-formyl-4-methoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OC)C=O)(O)O

Specifications

Melting Point ∽240°C (decomposition)
Quantity 5 g
Beilstein 7423998
Sensitivity Air Sensitive
Formula Weight 179.97
Percent Purity 98%
Chemical Name or Material 3-Formyl-4-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only