accessibility menu, dialog, popup

UserName

NH2SOHOO

CAS RN 121-57-3

IUPAC Name: 4-aminobenzene-1-sulfonic acid
Synonyms: sulfanilic acid kyselina sulfanilova sulfanilsaeure sodium salt 4-sulfanilic acid 4-sulfanilic acid, sodium salt 4-sulfanilic acid
Molecular Weight (g/mol): 173.19
Molecular Formula: C6H7NO3S
InChi Key: HVBSAKJJOYLTQU-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (10)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (5)
  • (3)
  • (2)
Grade 
  • (7)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (5)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (5)
  • (10)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
  • (5)