missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic Acid Standard, For Elementary Analysis, MilliporeSigma™

Catalog No. M1006840005
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1006840005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1006840005 Supplier Technidata Supplier No. 1.00684.0005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
PubChem CID 8479
ChEBI CHEBI:27500
IUPAC Name 4-aminobenzenesulfonic acid
SMILES C1=CC(=CC=C1N)S(=O)(=O)O

Specifications

Packaging Glass bottle
Quantity 5 g
Formula Weight 173.18. (molar mass)
Physical Form Solid
Chemical Name or Material Sulfanilic Acid Standard
Hazard Category H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.