missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic acid, 98+%

Catalog No. AAA1582530
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1582530 250 g
AAA158250B 1000 g
AAA158250I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1582530 Supplier Thermo Scientific Chemicals Supplier No. A1582530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent for the detection of histidine. Intermediate for dyes and pharmaceuticals | CAS: 121-57-3 | C6H7NO3S | 173.186 g/mol

Reagent for the detection of histidine; intermediate for dyes and pharmaceuticalsSulfanilic acid is used as a reagent for chromatographic detection of histidine. It is also used as a chemical intermediate in organic synthesis. It is utilized in quantitative analysis of nitrate and nitrite ions. It plays an essential role in the manufacturing of azo dyes and in synthesis of sulfa drugs. It is used as a dopant for the chemical synthesis of polyaniline. It is used to prepare 2,4-diamino-6-p-sulphoanilinopyrimidine by reacting with 6-chloro-pyrimidine-2,4-diamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for the detection of histidine; intermediate for dyes and pharmaceuticalsSulfanilic acid is used as a reagent for chromatographic detection of histidine. It is also used as a chemical intermediate in organic synthesis. It is utilized in quantitative analysis of nitrate and nitrite ions. It plays an essential role in the manufacturing of azo dyes and in synthesis of sulfa drugs. It is used as a dopant for the chemical synthesis of polyaniline. It is used to prepare 2,4-diamino-6-p-sulphoanilinopyrimidine by reacting with 6-chloro-pyrimidine-2,4-diamine.

Solubility
Slightly soluble in water. Insoluble in ethanol, ether, caustic soda and sodium carbonate.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00007886
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
PubChem CID 8479
ChEBI CHEBI:27500
IUPAC Name 4-aminobenzenesulfonic acid
SMILES C1=CC(=CC=C1N)S(=O)(=O)O

Specifications

Melting Point ∽365°C
Density 1.485
Assay Percent Range 98%
Quantity 250 g
Beilstein 908765
Merck Index 14,8926
Solubility Information Slightly soluble in water. Insoluble in ethanol,ether,caustic soda and sodium carbonate.
Formula Weight 173.19
Percent Purity ≥98%
Chemical Name or Material Sulfanilic Acid
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H317
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
EINECSNumber 204-482-5
RTECSNumber WP3895500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only