missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic Acid 99.0+%, TCI America™

Catalog No. S0120500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0120500G 500 g
S012025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0120500G Supplier TCI America Supplier No. S0120500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00007886
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
PubChem CID 8479
ChEBI CHEBI:27500
IUPAC Name 4-aminobenzenesulfonic acid
SMILES C1=CC(=CC=C1N)S(=O)(=O)O

Specifications

Melting Point 288°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 173.19
Percent Purity ≥99.0% (HPLC,T)
Chemical Name or Material Sulfanilic Acid
EINECSNumber (3)-1971
RTECSNumber WP3895500
TSCA Yes