accessibility menu, dialog, popup

UserName

CAS RN 121-02-8

IUPAC Name: 2-methyl-5-nitrobenzene-1-sulfonyl chloride
Synonyms: 2-methyl-5-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 235.64
Molecular Formula: C7H6ClNO4S
InChi Key: WPGVQDHXOUAJBW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1S(Cl)(=O)=O)[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)