accessibility menu, dialog, popup

UserName

CAS RN 12070-12-1

CW

CAS RN 12070-12-1

IUPAC Name: methanidylidynetungstenylium
Synonyms: tungsten carbide
Molecular Weight (g/mol): 195.85
Molecular Formula: CW
InChi Key: UONOETXJSWQNOL-UHFFFAOYSA-N
SMILES: [C-]#[W+]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)