CAS RN 12019-52-2
CAS RN 12019-52-2
IUPAC Name:
(cupriotellanyl)copper
Synonyms:
(cupriotellanyl)copper
Molecular Weight (g/mol):
254.69
Molecular Formula:
Cu2Te
InChi Key:
MZEWONGNQNXVKA-UHFFFAOYSA-N
SMILES:
[Cu][Te][Cu]
1
–
1
of
1
results
1
Copper(I) telluride, 99.5% (metals basis)
CAS: 12019-52-2 Molecular Formula: Cu2Te Molecular Weight (g/mol): 254.692 MDL Number: MFCD00049727 InChI Key: MZEWONGNQNXVKA-UHFFFAOYSA-N Synonym: copper telluride,cuprous telluride,copper i telluride,dicopper telluride,cuprous monotelluride,dicopper monotelluride,copper telluride cu0.67te0.33,cupriotellanyl copper PubChem CID: 6914517 IUPAC Name: copper;tellurium SMILES: [Cu].[Cu].[Te]