missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) telluride, 99.5% (metals basis)
Description
It is used as a colorent of glass and ceramics, rubber vulcanization accelerator, metallurgical additives, petrochemical catalyst, solar cells, thermoelectric refrigeration Materials, Infrared Detection Materials, Electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black-Blue |
| Physical Form | Pieces |
| Quantity | 25 g |
| Assay Percent Range | 99.5% (Metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 254.68 |
| Odor | Odorless |
| Refractive Index | 1.45 |
| Density | 7.27 |
| Chemical Name or Material | Copper(I) telluride |
Safety and Handling
| Hazard Category | H301-H332; oxic if swallowed. Harmful if inhaled. |
| Hazard Statement | Keep container in a well ventilated place. Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste. |
| EINECSNumber | 234-646-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only