accessibility menu, dialog, popup

UserName

CAS RN 1197922-04-5

IUPAC Name: 10',16'-bis({bis[3,5-bis(trifluoromethyl)phenyl](hydroxy)methyl})-4λ⁵-spiro[morpholine-4,13'-pentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosane]-1'(23'),2',4',6',8',10',15',17',19',21'-decaen-4-ylium bromide
Synonyms: 10',16'-bis({bis[3,5-bis(trifluoromethyl)phenyl](hydroxy)methyl})-4λ⁵-spiro[morpholine-4,13'-pentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosane]-1'(23'),2',4',6',8',10',15',17',19',21'-decaen-4-ylium bromide
Molecular Weight (g/mol): 1354.82
Molecular Formula: C60H36BrF24NO3
InChi Key: UVHZWWMJSAGSJH-UHFFFAOYSA-M
SMILES: [Br-].OC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=C2C[N+]3(CCOCC3)CC3=C(C=C4C=CC=CC4=C3C2=C2C=CC=CC2=C1)C(O)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results