CAS RN 119-47-1
IUPAC Name:
2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Synonyms:
cyanox 2246 methylene di-T-butylcresol
Molecular Weight (g/mol):
340.51
Molecular Formula:
C23H32O2
InChi Key:
KGRVJHAUYBGFFP-UHFFFAOYSA-N
SMILES:
CC1=CC(CC2=CC(C)=CC(=C2O)C(C)(C)C)=C(O)C(=C1)C(C)(C)C
1
–
1
of
1
results
1
2,2'-Methylenebis(6-tert-butyl-p-cresol) 99.0+%, TCI America™
CAS: 119-47-1 Molecular Formula: C23H32O2 Molecular Weight (g/mol): 340.51 MDL Number: MFCD00043641 InChI Key: KGRVJHAUYBGFFP-UHFFFAOYSA-N PubChem CID: 8398 IUPAC Name: 2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol SMILES: CC1=CC(CC2=CC(C)=CC(=C2O)C(C)(C)C)=C(O)C(=C1)C(C)(C)C