missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis(6-tert-butyl-p-cresol) 99.0+%, TCI America™

Catalog No. M021725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M021725G 25 g
M0217250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M021725G Supplier TCI America Supplier No. M021725G
Only null left

Chemical Identifiers

CAS 119-47-1
Molecular Formula C23H32O2
Molecular Weight (g/mol) 340.51
MDL Number MFCD00043641
InChI Key KGRVJHAUYBGFFP-UHFFFAOYSA-N
PubChem CID 8398
IUPAC Name 2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
SMILES CC1=CC(CC2=CC(C)=CC(=C2O)C(C)(C)C)=C(O)C(=C1)C(C)(C)C

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
Synonym Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 3,3′C-Di-tert-butyl-2,2′C-dihydroxy-5,5′C-dimethyldiphenylmethane, 2,2′C-Methylenebis(6-tert-butyl-4-methylphenol)
Formula Weight 340.51
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis(6-tert-butyl-p-cresol)
EINECSNumber (4)-0100
RTECSNumber PA3500000
TSCA Yes