CAS RN 118949-61-4
IUPAC Name:
2,6-bis[4-(propan-2-yl)-4,5-dihydro-1,3-oxazol-2-yl]pyridine
Synonyms:
2,6-bis(4-isopropyl-4,5-dihydro-1,3-oxazol-2-yl)pyridine
Molecular Weight (g/mol):
301.39
Molecular Formula:
C17H23N3O2
InChi Key:
CSGQGLBCAHGJDR-UHFFFAOYNA-N
SMILES:
CC(C)C1COC(=N1)C1=CC=CC(=N1)C1=NC(CO1)C(C)C
1
–
1
of
1
results
1
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™
CAS: 118949-61-4 Molecular Formula: C17H23N3O2 Molecular Weight (g/mol): 301.39 MDL Number: MFCD00191815 InChI Key: CSGQGLBCAHGJDR-HUUCEWRRSA-N Synonym: (S,S)-2,2′C-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline) PubChem CID: 688211 IUPAC Name: (4S)-4-propan-2-yl-2-[6-[(4S)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole SMILES: CC(C)C1COC(=N1)C2=NC(=CC=C2)C3=NC(CO3)C(C)C