missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™

Catalog No. B22185G
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22185G 5 g
B2218500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22185G Supplier TCI America Supplier No. B22185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118949-61-4
Molecular Formula C17H23N3O2
Molecular Weight (g/mol) 301.39
MDL Number MFCD00191815
InChI Key CSGQGLBCAHGJDR-HUUCEWRRSA-N
Synonym (S,S)-2,2′C-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)
PubChem CID 688211
IUPAC Name (4S)-4-propan-2-yl-2-[6-[(4S)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)C1COC(=N1)C2=NC(=CC=C2)C3=NC(CO3)C(C)C

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 5 g
Formula Weight 301.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine