accessibility menu, dialog, popup

UserName

HN

CAS RN 118864-75-8

IUPAC Name: 1-phenyl-1,2,3,4-tetrahydroisoquinoline
Synonyms: 1-phenyl-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol): 209.29
Molecular Formula: C15H15N
InChi Key: PRTRSEDVLBBFJZ-UHFFFAOYNA-N
SMILES: C1CC2=CC=CC=C2C(N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results