missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98.0+%, TCI America™

Catalog No. P22165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22165G 5 g
P22161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22165G Supplier TCI America Supplier No. P22165G

Chemical Identifiers

CAS 118864-75-8
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.292
MDL Number MFCD08692036
InChI Key PRTRSEDVLBBFJZ-HNNXBMFYSA-N
PubChem CID 1382087
IUPAC Name (1S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline
SMILES C1CNC(C2=CC=CC=C21)C3=CC=CC=C3

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Formula Weight 209.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline